8-Amino-5-methoxyquinoline
Synonym(s):
Gong Chen Auxiliary, MQ Auxiliary
Empirical Formula (Hill Notation):
C10H10N2O
CAS Number:
Molecular Weight:
174.20
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Showing 1-1 of 1