
7-Methylxanthine

Synonym(s):
7-Methylxanthine, 2,6-Dihydroxy-7-methylpurine, Heteroxanthine
Empirical Formula (Hill Notation):
C6H6N4O2
CAS Number:
Molecular Weight:
166.14
Products
Brand
Formula Weight
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing