
6´,-7´-Dihydroxybergamottin

Empirical Formula (Hill Notation):
C21H24O6
CAS Number:
Molecular Weight:
372.41
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Research Area
Showing 1-1 of 1
Compare
Product Number
Description
Pricing