
5-Vinyl-2-pyrrolidone

Synonym(s):
(5RS)-5-Ethenylpyrrolidin-2-one, 5-Vinyl-2-pyrrolidinone
Empirical Formula (Hill Notation):
C6H9NO
CAS Number:
Molecular Weight:
111.14
Products
Formula Weight
Manufacturer
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing