Skip to Content
Merck
4-Methyl-5-vinylthiazole ≥97%, FG

4-Methyl-5-vinylthiazole

4-Methyl-5-vinylthiazole ≥97%, FG
Empirical Formula (Hill Notation):
C6H7NS
CAS Number:
Molecular Weight:
125.19
EC Number:

Products

Product Category
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing