
4-Hydroxyestradiol

Synonym(s):
1,3,5(10)-Estratriene-3,4,17β-triol
Empirical Formula (Hill Notation):
C18H24O3
CAS Number:
Molecular Weight:
288.38
Products
Brand
Formula Weight
Gene Alias
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing