![4-[2-(tert-Butylamino)-1-methoxyethyl]-2-(hydroxymethyl)phenol certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/127/263/f2a2de27-a9eb-412b-bb4c-c9ec9329ceb7/640/f2a2de27-a9eb-412b-bb4c-c9ec9329ceb7.png)
4-[2-(tert-Butylamino)-1-methoxyethyl]-2-(hydroxymethyl)phenol
![4-[2-(tert-Butylamino)-1-methoxyethyl]-2-(hydroxymethyl)phenol certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/127/263/f2a2de27-a9eb-412b-bb4c-c9ec9329ceb7/640/f2a2de27-a9eb-412b-bb4c-c9ec9329ceb7.png)
Synonym(s):
5-[(1RS)-2-[(1,1-Dimethylethyl)amino]-1-methoxyethyl]-2-hydroxyphenyl]methanol, 5-[2-[(1,1-Dimethylethyl)amino]-1-methoxyethyl]-2-hydroxybenzenemethanol
Empirical Formula (Hill Notation):
C14H23NO3
CAS Number:
Molecular Weight:
253.34
Products
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing