3-Hydroxy-1,2-dimethyl-4(1H)-pyridone
Synonym(s):
Deferiprone
Empirical Formula (Hill Notation):
C7H9NO2
CAS Number:
Molecular Weight:
139.15
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1