
3-Fluoro-DL-valine

Synonym(s):
2-Amino-3-fluoro-3-methylbutyric acid
Empirical Formula (Hill Notation):
C5H10FNO2
CAS Number:
Molecular Weight:
135.14
Products
Product Category
Brand
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Amino Acid Type
Reaction Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing