
(3-Carboxypropyl)trimethylammonium chloride

Synonym(s):
γ-Butyrobetaine hydrochloride, Deoxycarnitine hydrochloride
Linear Formula:
C7H16NO2Cl
CAS Number:
Molecular Weight:
181.66
Products
Formula Weight
Melting Point (°C)
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing