
(2,4-Dichlorophenoxy)acetic acid sodium salt monohydrate

Synonym(s):
Sodium 2,4-dichlorophenoxyacetate monohydrate
Empirical Formula (Hill Notation):
C8H5Cl2NaO3 · H2O
CAS Number:
Molecular Weight:
261.03
Products
Brand
Formula Weight
Physical Form
Product Line
Application
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing