Skip to Content
Merck
2,2′-Bipyridyl ReagentPlus®, ≥99%

2,2′-Bipyridyl

2,2′-Bipyridyl ReagentPlus®, ≥99%
Synonym(s):
2,2′-Bipyridine, 2,2′-Dipyridine, 2,2′-Dipyridyl
Empirical Formula (Hill Notation):
C10H8N2
CAS Number:
Molecular Weight:
156.18
EC Number:

Products

Brand
Sustainability
Biological Source
Boiling Point (°C)
Feature
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Markush Class
Markush Group
Special Grade
Showing 1-2 of 2