
1α,25-Dihydroxyvitamin D3 (6,19,19-d3) solution

Synonym(s):
1α,25-Dihydroxycholecalciferol (6,19,19-d3) solution, Calcitriol (6,19,19-d3) solution, Deuterated 1-α25-Dihydroxyvitamin D3 solution
Empirical Formula (Hill Notation):
C27D3H41O3
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Quality Segment
Isotope
Application
Markush Class
Markush Group
Technique
Showing 1-2 of 2