
1,5-Bis(diphenylphosphino)pentane

Synonym(s):
dpppe
Linear Formula:
(C6H5)2P(CH2)5P(C6H5)2
CAS Number:
Molecular Weight:
440.50
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Reaction Type
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing