Merck
HomeChemical Vapor DepositionFew Monolayer Atomic Layer Deposition (ALD) on Surfaces and Interfaces for Energy Applications
Few Monolayer Atomic Layer Deposition (ALD) on Surfaces and Interfaces for Energy Applications
Introduction
Atomic layer deposition (ALD) exploits a sequential set of two or more surface reactions repeated cyclically to achieve thin film growth.1 These surface reactions occur between a chemisorbed species attached to a substrate and incoming precursor molecules from the gas-phase. Due to its precise deposition mechanism, ALD possesses many advantages over other thin film deposition processes. First, ALD offers extremely precise control at the angstrom level over the thickness of the deposited layer. Second, ultrahigh-aspect-ratio structures, including nanoporous solids2 and three-dimensional (3D) hierarchical structures,3 can be conformally and uniformly covered by ALD without thickness or composition gradients. Third, the chemical composition of films can be controlled by tailoring ALD cycles in a programmable manner.4,5 Finally, ALD enables deposition at lower temperatures,6 making it possible to use with polymer-based7 and biological substrates that are incompatible with higher temperatures.8
It is not widely appreciated that only a few monolayers of material added to a surface or interface can significantly impact performance, quality, and reliability. Since ALD allows atomic scale resolution in deposition of monolayers, new scientific mechanisms and phenomena can be investigated by characterizing these surfaces. While there are many excellent review articles on applied ALD research,9–17 none focus on the impact of few monolayer ALD films on materials and devices. Therefore, this review highlights research using few monolayer ALD to enhance material and device quality, performance, and reliability, specifically in the fields of photovoltaics, batteries, and catalysis.
We define “few monolayers” as less than 10 layers or a thickness of less than 2–3 nm. This demarcation of a ‘few’ is based on research reviews, which show impressive improvements in performance and reliability within this thickness range, and in most instances actual performance suppression by a thicker film. This definition also includes descriptions such as ultrathin, thin, compact, and few layer.
Photovoltaics
ALD has been used to suppress charge recombination at interfaces in photovoltaic devices such as hybrid organicinorganic perovskite solar cells, dye-sensitized solar cells (DSSCs), and quantum dot solar cells (QDSCs). Also, with the advent of hybrid organic-inorganic perovskite solar cells, ALD has been effectively used to improve the stability of these air sensitive compounds.
TiO2 (up to four nm) was deposited by ALD on spin-coated mesoporous nanoparticulate TiO2 films as blocking layers to make high efficiency solid-state perovskite-based (CH3NH3PbI3) solar cells.18 As shown in Figure 1A, a two nm conformal TiO2 ALD overlayer was sufficient to block the electron recombination, leading to a large open-circuit potential of 969.4 mV (reference potential 836.6 mV) and the high photon-to-current efficiency (PCE) of 11.5%.
Few layer ALD has been employed to enhance the PCE of DSSCs.19–22 An ultrathin TiO2 layer (10 cycles, 0.3-0.6 nm) was successfully coated on the surface of submicrometer-sized aggregates of ZnO nanocrystallites.19 As shown in Figure 1B, both the open-circuit voltage and the fill factor were increased by the introduction of a TiO2 layer, due to suppression of surface charge recombination, without impairing the photocurrent density. This resulted in a more than 20% enhancement of PCE from 5.2% to 6.3%.
Two nm ALD TiO2 on mesoporous TiO2
Figure 1. A) Two nm ALD TiO2 on mesoporous TiO2 is sufficient to block electron hole recombination in solid-state perovskite-based solar cells. Reproduced with permission from reference 18, copyright 2014 Wiley. B) 0.6 nm of ALD TiO2 on nanocrystalline ZnO suppresses charge recombination in DSSCs. Reproduced with permission from reference 19, copyright 2010 Wiley. C) 0.3 nm Al2O3 improves ambient stability of hybrid organic–inorganic perovskite solar cells to up to 24 days. Reproduced with permission from reference 24, copyright 2015 Royal Society of Chemisty.
Conformal TiO2 interfacial layers with a thickness of 1.5, 2.2, and 3.2 nm were coated on CdS quantum dots (QDs) and N719 dyes to enhance the performance of quantum dot and dye co-sensitized solar cells.23 The introduction of TiO2 interfacial layers significantly enhanced the short-circuit current density and PCE by improving the stability of the QDs and reducing the electron recombination with the electrolyte. The highest PCE reported was 2.36%, a 41% improvement from the reference device, at the optimal TiO2 thickness of 2.2 nm.
Ultrathin compact Al2O3 layers were introduced by ALD (1-7 cycles) onto a layer of hole-transport material (HTM) to improve the stability of hybrid organic–inorganic perovskite solar cells in humid conditions.24 The experimental results shown in Figure 1C indicate that a 3-cycle (~0.3 nm) ALD Al2O3 layer greatly improved the ambient stability of organic–inorganic perovskite solar cells and retained 90% of the initial PCE after 24 days of storage in air. When the number of ALD cycles increased, the current decreased sharply, probably caused by a significant reduction in the quantum tunneling of the holes.
Batteries
Batteries face limitations in energy storage density, charging rate, cycling stability, safety, and cost.17 Few layer ALD can effectively address many of these issues. For example, nanosized LiCoO2 powders were prepared by a molten salt method, and then coated directly using ALD (2 and 6 cycle) to make ultrathin conformal Al2O3 films (Figure 2A).25 The nano- LiCoO2 electrodes coated with 2 ALD cycles of Al2O3 (0.22 nm) delivered the highest discharge capacity of 133 mAh/g at a current rate of 1400 mA/g (7.8 C), corresponding to a 250% improvement in reversible capacity when compared to bare LiCoO2 nanoparticles. The improved performance resulted from the suppression of cobalt dissolution from the LiCoO2 by the ALD Al2O3 film. However, coating nano-LiCoO2 electrodes with 6 ALD cycles of Al2O3 decreased the rate performance, indicating that thick films resulted in poor Li+ conductivity.
Nanostructured anodes coated by few layer ALD have also shown enhanced rate performance and cycling stability.26 Porous Fe2O3 nanorods anchored on nitrogen-doped graphene sheets (NGr) (Figure 2B) are one example. ALD was used to deposit two cycles of Al2O3 film (<1 nm) on the Fe2O3 nanorods anchored on NGr (NGr-M-I-2ALD). This anode displayed a first-cycle Coulombic efficiency of 89%, much higher than uncoated NGr-I-M (65%). Further, the stable capacity of the anodes (249 mAh/g) at 20 A/g was maintained for 200 cycles.
Few layer ALD was also used for cathode modification to improve the performance of a lithium-sulfur (Li-S) battery.27 Al2O3 was deposited by ALD on a graphene-sulfur (G-S) composite electrode. The specific capacity (646 mAh/g) of a G-S composite cathode with 10 ALD cycles of Al2O3 film (~1 nm) after 100 charge/discharge cycles at 0.5 oC was 103% higher than that of bare G-S (Figure 2C). The coulombic efficiency, rate capability, and electrochemical stability of the G-S composite electrode were also significantly increased by ALD– Al2O3 coating.
Other examples of the use of few monolayer ALD in lithium ion batteries include direct deposition of Al2O3 on natural graphite particles (Figure 2D).28 These electrodes exhibit improved capacity retention when compared to bare electrodes because the insulating Al2O3 layer blocks electron conduction paths between natural graphite particles and the current collector. Ultrathin Al2O3 layers were coated directly on Li metal foils without air exposure to produce anodes for Li symmetric cells. The Li metal anodes treated with 20 ALD cycles of Al2O3 (~2 nm) were used for up to 1259 cycles before failure, double the lifetime of non-treated anodes (control sample = 711 cycles) (Figure 2E).29 Few layer ALD also helped prevent dendrite formation and improve the homogeneity of current distribution along the Li electrode/solid electrolyte interphase (SEI) interface.
0.2 nm Al2O3 coating on nanosized LiCoO2
Figure 2.A) 0.2 nm Al2O3 coating on nanosized LiCoO2 leads to increase capacity and stability. Reproduced with permission from reference 25, copyright 2011 American Chemical Society. B) High stability of 0.2 nm Al2O3 coated on Fe2O3 attached to N-doped graphene sheets (NGr-I-M-2ALD) maintained capacity for 200 cycles. Reproduced with permission from reference 26, copyright 2014 Elsevier. C) 1 nm Al2O3 deposited on graphene sulfur composite electrodes shows 103% high specific capacity compared to uncoated electrodes. Reproduced with permission from reference 27, copyright 2014 Royal Society of Chemistry. D) Direct deposition of Al2O3 on graphitic electrodes shows stable capacity behavior. Reproduced with permission from reference 28, copyright 2010 Wiley. Reproduced with permission from reference 29, copyright 2015 American Chemical Society. E) 2 nm Al2O3 on Li metal foils shows improved cyclability and prevents dendrite formation.
Catalysis
The challenges in catalyst development are improving catalytic activity, selectivity, and stability with minimal byproducts.14 Current synthetic methods, which include impregnation, ion exchange, and precipitation, are limited in facing these challenges because of the non-uniform distribution of surface sites and the relatively narrow set of starting materials available. Few monolayer ALD has been tested as a way to address these challenges.
Metal oxide catalysts with high catalytic activity and stability have been developed by few monolayer ALD.30 TiO2 thin films (15 nm) were deposited by ALD on supported Ag nanoparticles, while SiO2 thin interlayers with thicknesses of 2, 5, 10, and 20 nm were deposited by ALD to separate the TiO2 film from Ag nanoparticles. The photocatalytic activities of the samples and references were investigated by degradation of methylene blue (MB) in UV light illumination (Figure 3A). TiO2/SiO2/Ag architectures with a 2 nm SiO2 interlayer displayed the best photocatalytic performance due to blue-shifting in the localized surface plasmon resonance (LSPR) and coupling energy from Ag to TiO2 bandgap. Thicker SiO2 interlayers decreased the photocatalytic activity because they reduced the penetration depth of the LSPR electromagnetic field into the SiO2 layer.
Few layer ALD was also employed for the photocatalytic conversion of CO2 to CO.31 Dense ZnO islands with different thickness were coated via ALD onto large arrays of massively parallel, single crystalline, high-density CuO nanowires generated by thermal oxidation of high-purity copper foils. The CO yield first increased and then decreased with increasing ALD cycles, and CuO nanowires with eight ALD cycles of ZnO islands (1.4 nm) displayed the optimal performance (Figure 3B). This behavior occurred because 1) the favorable band alignment and epitaxial matching between ALD ZnO nanoislands and CuO nanowires led to recombination-free charge transfer, 2) the surface morphology allowed gas exposure to both ZnO and CuO surfaces, and 3) surface defects on exposed ZnO islands resulted in lower quantum yields due to the trapping of transferred electrons.
The catalytic activity, selectivity, and stability of supported metal catalysts were also improved by few layer ALD.32–34 One of the most important catalysts is supported platinum. Platinum nanoparticles were deposited on Al2O3 by ALD (one or five cycles) with H2 or O2 as the second half-cycle precursor for oxidative dehydrogenation of propane. Using H2 as the second precursor resulted in smaller Pt particles with lower average coordination numbers than O2. After one ALD cycle (1.1 nm, measured by TEM), Pt catalyst with H2 as the second precursor exhibited the highest C3H6 selectivity (37% at 14% conversion at 400 ˚C), likely due to the small particle size (Figure 3C). Commercial Pt catalysts (3.9 nm, measured by TEM) had a selectivity of less than 1% at 9% conversion under the same reaction conditions.32
TiO2 overcoated Co/TiO2 catalysts were also prepared to replace precious-metal catalysts, like Pt and Ru, for aqueous-phase hydrogenation (APH) reactions.35 Generally, base metals (like cobalt) are inexpensive, abundant, and active for liquid phase reactions, but they have low stability caused by irreversible deactivation. Figure 3D shows that a thin ALD TiO2 coating prevented both leaching and sintering of Co particles during aqueous-phase reactions, due to TiO2 decoration of undercoordinated cobalt sites located at defects, corners, and edges. The regenerated TiO2/Co/TiO2 catalysts (30 ALD cycles, 1.2 nm) displayed much higher catalytic reactivity and stability than fresh catalysts for APH of furfuryl alcohol because more Co active sites were exposed after regeneration.35
 Fast decay in methylene blue
Figure 3. A) Fast decay in methylene blue from TiO2/SiO2/Ag with 2 nm SiO2 separation layer (green decay curve). Reproduced with permission from reference 30, copyright 2011 American Chemial Society. B) ZnO islands on CuO nanowire have optimal size (1.4 nm) for maximal CO2 → CO conversion. Reproduced with permission from reference 31, copyright 2015 American Chemial Society. C) Pt nanoparticle catalyst (1.1 nm) using H2 as reactant shows highest selectivity to oxidative dehydrogenation to propane at 400 ˚C. Reproduced with permission from reference 32, copyright 2015 American Chemial Society. D) 1.2 nm TiO2 on Co on TiO2 showed highest activity and stability for aqueous phase hydrogenation reactions. Reproduced with permission from reference 35, copyright 2014 Royal Society of Chemisty.
Conclusions
In summary, ALD is a singularly powerful technique that can be applied to tailor surfaces and interfaces using few monolayer films, which have a significant impact on heterogeneous material and device performance. The structure of the few monolayers can be precisely engineered, depending on how the precursor molecule interacts with the chemisorbed ligands on the substrate. If the deposition is layer-by-layer, conformal films are produced that are ideal for charge blocking or for providing surface passivation and protection. ALD films using these methods have been applied in a number of photovoltaics and batteries. On the other hand, if the ALD chemistry is driven by island nucleation followed by growth, then surfaces and interfaces can be decorated with nanocrystals in order to store charge and present chemically active facets useful for inducing chemical reactions. Their diversity in tailoring surfaces and interfaces is unique to ALD processes.
As ALD processes continue to develop, new precursor molecules are needed to provide greater flexibility for tailoring surfaces and interfaces. The interaction of precursor ligands with substrate functional groups controls nucleation of ALD films at cycle ‘one’. However, it is only now becoming clear that the interaction of nearest-neighbor adsorbed precursor molecules can affect film composition, structure, and thus properties.36–43 This raises the interesting question as to whether the design of precursor molecules with appropriate ligand chemistry can be exploited to deterministically control film nucleation and growth on any surface.
Regardless of the application, ALD at its ultimate thickness limit has heralded a new era of surface and interface engineering. The control of this process appears to be remarkably simple, and yet has hidden complexities that will continue to push the boundaries of discovery of new materials and concept devices, and allow even finer control over their properties and applications.
Acknowledgements
The authors acknowledge financial and material support from EMD Performance Materials. Helpful discussions with Dr. Ravi Kanjolia, Dr. Charles Dezelah and Dr. Jacob Woodruff are acknowledged. Partial funding was provided by U.S. Army RDECOM Acquisition Grant W911NF-15-1-0178, Subgrant RSC15032. Partial funding was also provided through the U.S.- India Partnership to Advance Clean Energy-Research (PACE-R) for the Solar Energy Research Institute for India and the United States (SERIIUS), funded by the U.S. Department of Energy (Office of Science, Office of Basic Energy Sciences, and Energy Efficiency and Renewable Energy, Solar Energy Technology Program, under subcontract DE-AC36-08GO28308 to the national Renewable Energy Laboratory, Golden, Colorado).
Materials
Sorry, an unexpected error has occurred
Network error: Failed to fetch
References
1.
George SM. 2010. Atomic Layer Deposition: An Overview. Chem. Rev.. 110(1):111-131. https://doi.org/10.1021/cr900056b
2.
Banerjee P, Perez I, Henn-Lecordier L, Lee SB, Rubloff GW. 2009. Nanotubular metal?insulator?metal capacitor arrays for energy storage. Nature Nanotech. 4(5):292-296. https://doi.org/10.1038/nnano.2009.37
3.
Bielinski AR, Boban M, He Y, Kazyak E, Lee DH, Wang C, Tuteja A, Dasgupta NP. 2017. Rational Design of Hyperbranched Nanowire Systems for Tunable Superomniphobic Surfaces Enabled by Atomic Layer Deposition. ACS Nano. 11(1):478-489. https://doi.org/10.1021/acsnano.6b06463
4.
Banerjee P, Lee W, Bae K, Lee SB, Rubloff GW. 2010. Structural, electrical, and optical properties of atomic layer deposition Al-doped ZnO films. Journal of Applied Physics. 108(4):043504. https://doi.org/10.1063/1.3466987
5.
Gao Z, Myung Y, Huang X, Kanjolia R, Park J, Mishra R, Banerjee P. 2016. Doping Mechanism in Transparent, Conducting Tantalum Doped ZnO Films Deposited Using Atomic Layer Deposition. Adv. Mater. Interfaces. 3(21):1600496. https://doi.org/10.1002/admi.201600496
6.
Groner MD, Fabreguette FH, Elam JW, George SM. 2004. Low-Temperature Al2O3Atomic Layer Deposition. Chem. Mater.. 16(4):639-645. https://doi.org/10.1021/cm0304546
7.
Parsons GN, Atanasov SE, Dandley EC, Devine CK, Gong B, Jur JS, Lee K, Oldham CJ, Peng Q, Spagnola JC, et al. 2013. Mechanisms and reactions during atomic layer deposition on polymers. Coordination Chemistry Reviews. 257(23-24):3323-3331. https://doi.org/10.1016/j.ccr.2013.07.001
8.
Knez M, Kadri A, Wege C, Gösele U, Jeske H, Nielsch K. 2006. Atomic Layer Deposition on Biological Macromolecules:  Metal Oxide Coating of Tobacco Mosaic Virus and Ferritin. Nano Lett.. 6(6):1172-1177. https://doi.org/10.1021/nl060413j
9.
Knez M, Nielsch K, Niinistö L. 2007. Synthesis and Surface Engineering of Complex Nanostructures by Atomic Layer Deposition. Adv. Mater.. 19(21):3425-3438. https://doi.org/10.1002/adma.200700079
10.
Kim H, Lee H, Maeng W. 2009. Applications of atomic layer deposition to nanofabrication and emerging nanodevices. Thin Solid Films. 517(8):2563-2580. https://doi.org/10.1016/j.tsf.2008.09.007
11.
Parsons GN, George SM, Knez M. 2011. Progress and future directions for atomic layer deposition and ALD-based chemistry. MRS Bull.. 36(11):865-871. https://doi.org/10.1557/mrs.2011.238
12.
Elam JW, Dasgupta NP, Prinz FB. 2011. ALD for clean energy conversion, utilization, and storage. MRS Bull.. 36(11):899-906. https://doi.org/10.1557/mrs.2011.265
13.
Johnson RW, Hultqvist A, Bent SF. 2014. A brief review of atomic layer deposition: from fundamentals to applications. Materials Today. 17(5):236-246. https://doi.org/10.1016/j.mattod.2014.04.026
14.
O'Neill BJ, Jackson DHK, Lee J, Canlas C, Stair PC, Marshall CL, Elam JW, Kuech TF, Dumesic JA, Huber GW. 2015. Catalyst Design with Atomic Layer Deposition. ACS Catal.. 5(3):1804-1825. https://doi.org/10.1021/cs501862h
15.
Dasgupta NP, Lee H, Bent SF, Weiss PS. 2016. Recent Advances in Atomic Layer Deposition. Chem. Mater.. 28(7):1943-1947. https://doi.org/10.1021/acs.chemmater.6b00673
16.
Dasgupta NP, Li L, Sun X. 2016. Atomic Layer Deposition for Energy and Environmental Applications. Adv. Mater. Interfaces. 3(21): https://doi.org/10.1002/admi.201600914
17.
Meng X, Wang X, Geng D, Ozgit-Akgun C, Schneider N, Elam JW. Atomic layer deposition for nanomaterial synthesis and functionalization in energy technology. Mater. Horiz.. 4(2):133-154. https://doi.org/10.1039/c6mh00521g
18.
Chandiran AK, Yella A, Mayer MT, Gao P, Nazeeruddin MK, Grätzel M. 2014. Sub-Nanometer Conformal TiO2Blocking Layer for High Efficiency Solid-State Perovskite Absorber Solar Cells. Adv. Mater.. 26(25):4309-4312. https://doi.org/10.1002/adma.201306271
19.
Park K, Zhang Q, Garcia BB, Zhou X, Jeong Y, Cao G. 2010. Effect of an Ultrathin TiO2 Layer Coated on Submicrometer-Sized ZnO Nanocrystallite Aggregates by Atomic Layer Deposition on the Performance of Dye-Sensitized Solar Cells. Adv. Mater.. 22(21):2329-2332. https://doi.org/10.1002/adma.200903219
20.
Shanmugam M, Baroughi MF, Galipeau D. 2010. Effect of atomic layer deposited ultra thin HfO2 and Al2O3 interfacial layers on the performance of dye sensitized solar cells. Thin Solid Films. 518(10):2678-2682. https://doi.org/10.1016/j.tsf.2009.08.033
21.
Chandiran AK, Tetreault N, Humphry-Baker R, Kessler F, Baranoff E, Yi C, Nazeeruddin MK, Grätzel M. 2012. Subnanometer Ga2O3 Tunnelling Layer by Atomic Layer Deposition to Achieve 1.1 V Open-Circuit Potential in Dye-Sensitized Solar Cells. Nano Lett.. 12(8):3941-3947. https://doi.org/10.1021/nl301023r
22.
Alibabaei L, Farnum BH, Kalanyan B, Brennaman MK, Losego MD, Parsons GN, Meyer TJ. 2014. Atomic Layer Deposition of TiO2on Mesoporous nanoITO: Conductive Core?Shell Photoanodes for Dye-Sensitized Solar Cells. Nano Lett.. 14(6):3255-3261. https://doi.org/10.1021/nl5006433
23.
Lin X, Yu K, Lu G, Chen J, Yuan C. 2013. Atomic layer deposition of TiO2interfacial layer for enhancing performance of quantum dot and dye co-sensitized solar cells. J. Phys. D: Appl. Phys.. 46(2):024004. https://doi.org/10.1088/0022-3727/46/2/024004
24.
Dong X, Fang X, Lv M, Lin B, Zhang S, Ding J, Yuan N. Improvement of the humidity stability of organic?inorganic perovskite solar cells using ultrathin Al2O3layers prepared by atomic layer deposition. J. Mater. Chem. A. 3(10):5360-5367. https://doi.org/10.1039/c4ta06128d
25.
Scott ID, Jung YS, Cavanagh AS, Yan Y, Dillon AC, George SM, Lee S. 2011. Ultrathin Coatings on Nano-LiCoO2for Li-Ion Vehicular Applications. Nano Lett.. 11(2):414-418. https://doi.org/10.1021/nl1030198
26.
Hu T, Xie M, Zhong J, Sun H, Sun X, Scott S, George SM, Liu C, Lian J. 2014. Porous Fe2O3 nanorods anchored on nitrogen-doped graphenes and ultrathin Al2O3 coating by atomic layer deposition for long-lived lithium ion battery anode. Carbon. 76141-147. https://doi.org/10.1016/j.carbon.2014.04.060
27.
Yu M, Yuan W, Li C, Hong J, Shi G. 2014. Performance enhancement of a graphene?sulfur composite as a lithium?sulfur battery electrode by coating with an ultrathin Al2O3 film via atomic layer deposition. J. Mater. Chem. A. 2(20):7360. https://doi.org/10.1039/c4ta00234b
28.
Jung YS, Cavanagh AS, Riley LA, Kang S, Dillon AC, Groner MD, George SM, Lee S. Ultrathin Direct Atomic Layer Deposition on Composite Electrodes for Highly Durable and Safe Li-Ion Batteries. Adv. Mater.. 22(19):2172-2176. https://doi.org/10.1002/adma.200903951
29.
Kazyak E, Wood KN, Dasgupta NP. 2015. Improved Cycle Life and Stability of Lithium Metal Anodes through Ultrathin Atomic Layer Deposition Surface Treatments. Chem. Mater.. 27(18):6457-6462. https://doi.org/10.1021/acs.chemmater.5b02789
30.
Kumar MK, Krishnamoorthy S, Tan LK, Chiam SY, Tripathy S, Gao H. 2011. Field Effects in Plasmonic Photocatalyst by Precise SiO2 Thickness Control Using Atomic Layer Deposition. ACS Catal.. 1(4):300-308. https://doi.org/10.1021/cs100117v
31.
Wang W, Wu F, Myung Y, Niedzwiedzki DM, Im HS, Park J, Banerjee P, Biswas P. 2015. Surface Engineered CuO Nanowires with ZnO Islands for CO2Photoreduction. ACS Appl. Mater. Interfaces. 7(10):5685-5692. https://doi.org/10.1021/am508590j
32.
Gould TD, Lubers AM, Corpuz AR, Weimer AW, Falconer JL, Medlin JW. 2015. Controlling Nanoscale Properties of Supported Platinum Catalysts through Atomic Layer Deposition. ACS Catal.. 5(2):1344-1352. https://doi.org/10.1021/cs501265b
33.
Yan H, Cheng H, Yi H, Lin Y, Yao T, Wang C, Li J, Wei S, Lu J. 2015. Single-Atom Pd1/Graphene Catalyst Achieved by Atomic Layer Deposition: Remarkable Performance in Selective Hydrogenation of 1,3-Butadiene. J. Am. Chem. Soc.. 137(33):10484-10487. https://doi.org/10.1021/jacs.5b06485
34.
Wang H, Wang C, Yan H, Yi H, Lu J. 2015. Precisely-controlled synthesis of Au@Pd core?shell bimetallic catalyst via atomic layer deposition for selective oxidation of benzyl alcohol. Journal of Catalysis. 32459-68. https://doi.org/10.1016/j.jcat.2015.01.019
35.
Lee J, Jackson DHK, Li T, Winans RE, Dumesic JA, Kuech TF, Huber GW. 2014. Enhanced stability of cobalt catalysts by atomic layer deposition for aqueous-phase reactions. Energy Environ. Sci.. 7(5):1657. https://doi.org/10.1039/c4ee00379a
36.
Shi L, Gao Z, Liu Z, Myung Y, Banerjee P. 2017. Configurational Entropy of Adlayers in Atomic Layer Deposition. Chem. Mater.. 29(13):5458-5462. https://doi.org/10.1021/acs.chemmater.6b04829
37.
Shirazi M, Elliott SD. 2014. Atomistic kinetic Monte Carlo study of atomic layer deposition derived from density functional theory. J. Comput. Chem.. 35(3):244-259. https://doi.org/10.1002/jcc.23491
38.
Tanskanen JT, Hägglund C, Bent SF. 2014. Correlating Growth Characteristics in Atomic Layer Deposition with Precursor Molecular Structure: The Case of Zinc Tin Oxide. Chem. Mater.. 26(9):2795-2802. https://doi.org/10.1021/cm403913r
39.
Shirazi M, Elliott SD. Cooperation between adsorbates accounts for the activation of atomic layer deposition reactions. Nanoscale. 7(14):6311-6318. https://doi.org/10.1039/c5nr00900f
40.
Weckman T, Laasonen K. First principles study of the atomic layer deposition of alumina by TMA?H2O-process. Phys. Chem. Chem. Phys.. 17(26):17322-17334. https://doi.org/10.1039/c5cp01912e
41.
Maimaiti Y, Elliott SD. 2016. Kinetics and Coverage Dependent Reaction Mechanisms of the Copper Atomic Layer Deposition from Copper Dimethylamino-2-propoxide and Diethylzinc. Chem. Mater.. 28(17):6282-6295. https://doi.org/10.1021/acs.chemmater.6b02522
42.
Goehry C, Schneider N. 2017. Chemical Processes Involved in Atomic Layer Deposition of Gallium Sulfide: Insights from Theory. J. Phys. Chem. C. 121(11):5871-5881. https://doi.org/10.1021/acs.jpcc.6b12242
43.
Guerra-Nuñez C, Döbeli M, Michler J, Utke I. 2017. Reaction and Growth Mechanisms in Al2O3 deposited via Atomic Layer Deposition: Elucidating the Hydrogen Source. Chem. Mater.. 29(20):8690-8703. https://doi.org/10.1021/acs.chemmater.7b02759