Merck
HPLC Analysis of Sugars on SUPELCOGEL Ca
HPLC Analysis of Sugars on SUPELCOGEL™ Ca application for HPLC
Materials
analytical column
Product No.
Description
Pricing
SUPELCOGEL Ca, 6% Crosslinked HPLC Column
9 μm particle size, L × I.D. 30 cm × 7.8 mm
CONDITIONS
column
SUPELCOGEL Ca, 30 cm × 7.8 mm I.D., 9 μm particles (59305-U)
mobile phase
water
flow rate
1.5 mL/min
column temp.
80 °C
detector
RI
injection
10 μL, 2.5 mg/mL each analyte in water
Description
Legal Information
SUPELCOGEL is a trademark of Sigma-Aldrich Co. LLC