Merck
HPLC Analysis of Warfarin Enantiomers on Astec® CHIROBIOTIC® V (Method 2)
HPLC Analysis of Warfarin Enantiomers on Astec® CHIROBIOTIC® V (Method 2) application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Astec® CHIROBIOTIC® V Chiral HPLC Column
5 μm particle size, L × I.D. 25 cm × 4.6 mm
Application
Product No.
Description
Pricing
HPLC Analysis of Warfarin Anticoagulant on Ascentis® C18
application for HPLC
HPLC Analysis of Warfarin Enantiomers on Astec® CYCLOBOND I 2000 (UV at 254 nm)
application for HPLC
HPLC Analysis of Warfarin Enantiomers on Astec® CHIROBIOTIC® V2
application for HPLC
HPLC Analysis of Warfarin Enantiomers on Astec® Cellulose DMP
application for HPLC
standard
Product No.
Description
Pricing
Warfarin
analytical standard
Warfarin
PESTANAL®, analytical standard
(R)-(+)-Warfarin
≥97% (HPLC)
(S)-(−)-Warfarin
≥97% (HPLC)
CONDITIONS
column
CHIROBIOTIC V, 25 cm x 4.6 mm I.D., 5 μm particles (11024AST)
mobile phase
[A] 0.1% triethylamine acetate, pH 5.0; [B] THF; (80:20, A:B)
flow rate
1 mL/min
column temp.
ambient
detector
UV, 254 nm
injection
1 μL
sample
4 mg/mL in methanol
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC
null