HPLC Analysis of Sibutramine Enantiomers on Astec® CHIROBIOTIC® V2
![HPLC Analysis of Sibutramine Enantiomers on Astec® CHIROBIOTIC® V2 application for HPLC](/deepweb/assets/sigmaaldrich/product/images/242/076/37f762ec-c0f3-467f-857b-7b5dfb11e6f5/800/37f762ec-c0f3-467f-857b-7b5dfb11e6f5.jpg)
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
CHIROBIOTIC V2, 25 cm x 4.6 mm I.D., 5 μm particles (15024AST)
mobile phase
15 mM ammonium formate in methanol
flow rate
1 mL/min
column temp.
25 °C
detector
UV, 230 nm
injection
10 μL
sample
1 mg/mL in methanol
Description
Analysis Note
Sibutramine, one of a few drugs approved for long-term treatment of obesity, is a chiral compound. The (R) enantiomer has the desired effect of reducing body weight and food intake, but the (S) enantiomer has the opposite effect: A perfect example of why chiral HPLC is important to pharmaceutical and clinical analysts. Astec® CHIROBIOTIC® V2 successfully separates sibutramine enantiomers, using a simple, MS-friendly mobile phase.
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC