HPLC Analysis of Dorzolamide Enantiomers on Astec® CHIROBIOTIC® V2
![HPLC Analysis of Dorzolamide Enantiomers on Astec® CHIROBIOTIC® V2 application for HPLC](/deepweb/assets/sigmaaldrich/product/images/141/286/c02f19dc-c936-4451-92fc-7e1b2161e5d1/800/c02f19dc-c936-4451-92fc-7e1b2161e5d1.jpg)
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
CHIROBIOTIC V2, 25 cm x 4.6 mm I.D., 5 μm particles (15024AST)
injection
10 μL
mobile phase
[A] 3.81 mM ammonium trifluoroacetic acid; [B] methanol; (5:95, A:B)
flow rate
0.3 mL/min
column temp.
22 °C
detector
UV, 254 nm
sample
1 mg/mL in methanol
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC