HPLC Analysis of Bilirubin and Biliverdin on Discovery® C18
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
Discovery C18, 10 cm × 2.1 mm I.D., 5 μm particles (569220-U)
mobile phase
[A] 10 mM ammonium acetate (pH unadjusted) [B] 90:10, methanol:(100 mM ammonium acetate)
flow rate
0.2 mL/min
column temp.
ambient
detector
UV, 405 nm
injection
1 μL
sample
as indicated (in 50% MeOH, 1% NH4OH)
Description
Legal Information
Discovery is a registered trademark of Merck KGaA, Darmstadt, Germany