HPLC Analysis of 3-Methylaspartic Acid Enantiomers on Astec® CHIROBIOTIC® T
![HPLC Analysis of 3-Methylaspartic Acid Enantiomers on Astec® CHIROBIOTIC® T application for HPLC](/deepweb/assets/sigmaaldrich/product/images/281/162/8b5bdad4-4531-46cf-b760-a66719300e57/800/8b5bdad4-4531-46cf-b760-a66719300e57.jpg)
CONDITIONS
column
CHIROBIOTIC T, 25 cm x 4.6 mm I.D., 5 μm particles (12024AST)
mobile phase
[A] methanol; [B] water; [C] formic acid; (80:20:0.02, A:B:C)
flow rate
0.9 mL/min
column temp.
25 °C
detector
ELSD
injection
4 μL
sample
5 mg/mL in methanol
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC