Merck
US EPA Method 8015 (modified): GC Analysis of Gasoline Range Organics (GRO) on Equity-1 after Purge & Trap using "M" Trap
US EPA Method 8015 (modified): GC Analysis of Gasoline Range Organics (GRO) on Equity-1 after Purge & Trap using "M" Trap suitable for GC
Materials
analytical column
Product No.
Description
Pricing
Equity-1 Capillary GC Column
L × I.D. 30 m × 0.32 mm, df 1.00 μm
SLB®-5ms Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 0.25 μm
SLB®-5ms Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 0.50 μm
SLB®-5ms Capillary GC Column
L × I.D. 20 m × 0.18 mm, df 0.18 μm
accessory
Product No.
Description
Pricing
Wheaton wide-mouth bottle
capacity 32 oz (1000 mL), amber, white polypropylene cap, PTFE flat liner
Supeltex M-2B Double-Tapered Ferrule
I.D. 0.4 mm, 0.10-0.25 mm to like I.D. Column, pkg of 2 ea
Capillary Column Butt Connector
body only (ferrules not included)
guard column
Product No.
Description
Pricing
Intermediate Polar Fused Silica Tubing
L × I.D. 5.0 m × 0.25 mm
standard
Product No.
Description
Pricing
N-Nitrosodiphenylamine solution
certified reference material, 5000 μg/mL in methanol
Perfluorotributylamine (PFTBA)
analytical standard
EPA 8270 GC-MS Tuning Solution II
certified reference material, 1000 μg/mL each component in dichloromethane
EPA CLP Semivolatile Calibration Mix
certified reference material, 1000 μg/mL each component in dichloromethane : benzene (3:1)
EPA CLP Semivolatiles Surrogate Standards Mix
certified reference material, 4000 μg/mL each component in methylene chloride
EPA 8270 Semivolatile Internal Standard Mix
certified reference material, TraceCERT®, 2000 μg/mL each component in methylene chloride, ampule of 2 × 1 mL
EPA 8270 Benzidines Mix
certified reference material, TraceCERT®, 2000 μg/mL each component in methanol, ampule of 1 mL
CONDITIONS
sample/matrix
Each analyte at 50 ppb in 5 mL of water
purge trap
MOD BTEXTRAP M, Tekmar® 3000
purge
40 mL/min at 25 °C for 11 min
dry purge
2 min
desorption preheat
220 °C
bake
260 °C for 10 min
transfer line/valve temp.
110 °C
column
Equity-1, 30 m x 0.32 mm I.D., 1.0 μm (28057-U)
oven
40 °C (2 min), 5 °C/min to 75 °C, 10 °C/min to 200 °C
inj. temp.
200 °C
detector
FID, 250 °C
carrier gas
helium, 1.4 mL/min constant
injection
20:1 split
liner
0.75 mm I.D. SPME
Description
Legal Information
null
Tekmar is a registered trademark of Tekmar Co.