Merck
GC Analysis of Bergamot Essential Oil on SLB-5ms (10 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis
GC Analysis of Bergamot Essential Oil on SLB-5ms (10 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis suitable for GC
Materials
analytical column
Product No.
Description
Pricing
SLB®-5ms Capillary GC Column
L × I.D. 10 m × 0.10 mm, df 0.10 μm
CONDITIONS
column
SLB-5ms, 10 m x 0.10 mm I.D., 0.10 μm (28465-U)
oven
50 °C, 14.0 °C/min to 250 °C
inj. temp.
300 °C
detector
FID, 300 °C
carrier gas
hydrogen, 57.0 cm/sec, 184.2 kPa
injection
0.4 μL, 600:1 split
liner
0.75 mm ID
sample
bergamot oil, diluted 10:1 in hexane