Merck
  • [Simultaneous determination of pesticide residues in crops by liquid chromatography with tandem mass spectrometry].
[Simultaneous determination of pesticide residues in crops by liquid chromatography with tandem mass spectrometry].
Shokuhin eiseigaku zasshi. Journal of the Food Hygienic Society of Japan (2006-11-30)
Rie Ishii, Kunihiko Takahashi, Masakazu Horie
ABSTRACT
A simultaneous method using liquid chromatography coupled with electrospray ionization tandem mass spectrometry (LC-ESI-MS/MS) was developed for the determination of pesticide residues in crops. Mass spectral acquisition was performed in the positive mode by applying multiple reaction monitoring. In LC separation, an Atlantis dC18 Column was used with acetic acid-ammonium acetate-acetonitrile as the mobile phase. Pesticide residues in crops were extracted with acetone, and cleaned up by liquid-liquid separation with saturated salt solution and hexane, followed by an ENVI-Carb cartridge. The quantification limits of compounds in crops were below 5 ng/g. Eighty compounds were obtained with recoveries ranging from 60 to 130% at the level of 50 ng/g with RSD (%) of less than 15%. Fifty crop samples were analyzed by the developed method. Seven pesticide residues were detected in nine crops.
Product Description
Supelco
Supel QuE, PSA/C18/ENVI-Carb Tube, pack of 100, AOAC® 2007.01
Supelco
Supel QuE, PSA/ENVI-Carb Tube 2, pk of 50, suitable for EN 15662:2008 per BS, centrifuge tube volume 15 mL , Shaker Compatible
Supelco
Supel QuE, PSA/C18/ENVI-Carb Tube, pk of 50, AOAC® 2007.01, centrifuge tube volume 15 mL , Shaker Compatible
Supelco
Supel QuE, PSA/ENVI-Carb (EN) Cleanup Tube 1, pack of 100, suitable for EN 15662:2008 per BS
Supelco
Supel QuE, PSA/ENVI-Carb (EN) Cleanup Tube 2, pack of 100, suitable for EN 15662:2008 per BS
Supelco
Supel QuE, PSA/ENVI-Carb Tube 1, pk of 50, suitable for EN 15662:2008 per BS, centrifuge tube volume 15 mL , Shaker Compatible