Merck
  • Dynamic all-optical drug screening on cardiac voltage-gated ion channels.
Dynamic all-optical drug screening on cardiac voltage-gated ion channels.
Scientific reports (2018-01-20)
Jonas Streit, Sonja Kleinlogel
ABSTRACT
Voltage-gated ion channels (VGCs) are prime targets for the pharmaceutical industry, but drug profiling on VGCs is challenging, since drug interactions are confined to specific conformational channel states mediated by changes in transmembrane potential. Here we combined various optogenetic tools to develop dynamic, high-throughput drug profiling assays with defined light-step protocols to interrogate VGC states on a millisecond timescale. We show that such light-induced electrophysiology (LiEp) yields high-quality pharmacological data with exceptional screening windows for drugs acting on the major cardiac VGCs, including hNa