Merck
  • Structure-Based Kinase Profiling To Understand the Polypharmacological Behavior of Therapeutic Molecules.
Structure-Based Kinase Profiling To Understand the Polypharmacological Behavior of Therapeutic Molecules.
Journal of chemical information and modeling (2017-12-16)
Devawati Dutta, Ranjita Das, Chhabinath Mandal, Chitra Mandal
ABSTRACT
Several drugs elicit their therapeutic efficacy by modulating multiple cellular targets and possess varied polypharmacological actions. The identification of the molecular targets of a potent bioactive molecule is essential in determining its overall polypharmacological profile. Experimental procedures are expensive and time-consuming. Therefore, computational approaches are actively implemented in rational drug discovery. Here, we demonstrate a computational pipeline, based on reverse virtual screening technique using several consensus scoring strategies, and perform structure-based kinase profiling of 12 FDA-approved drugs. This target prediction showed an overall good performance, with an average AU-ROC greater than 0.85 for most drugs, and identified the true targets even at the top 2% cutoff. In contrast, 10 non-kinase binder drugs exhibited lower binding efficiency and appeared in the bottom of ranking list. Subsequently, we validated this pipeline on a potent therapeutic molecule, mahanine, whose polypharmacological profile related to targeting kinases is unknown. Our target-prediction method identified different kinases. Furthermore, we have experimentally validated that mahanine is able to modulate multiple kinases that are involved in cross-talk with different signaling molecules, which thereby exhibits its polypharmacological action. More importantly, in vitro kinase assay exhibited the inhibitory effect of mahanine on two such predicted kinases' (mTOR and VEGFR2) activity, with IC
Sigma-Aldrich
VEGFR2 human, recombinant, expressed in insect cells, ≥95% (SDS-PAGE)
Supelco
Malachite Green chloride, analytical standard