Merck
  • Covalent attachment of the heme to Synechococcus hemoglobin alters its reactivity toward nitric oxide.
Covalent attachment of the heme to Synechococcus hemoglobin alters its reactivity toward nitric oxide.
Journal of inorganic biochemistry (2017-10-03)
Matthew R Preimesberger, Eric A Johnson, Dillon B Nye, Juliette T J Lecomte
ABSTRACT
The cyanobacterium Synechococcus sp. PCC 7002 produces a monomeric hemoglobin (GlbN) implicated in the detoxification of reactive nitrogen and oxygen species. GlbN contains a b heme, which can be modified under certain reducing conditions. The modified protein (GlbN-A) has one heme-histidine C-N linkage similar to the C-S linkage of cytochrome c. No clear functional role has been assigned to this modification. Here, optical absorbance and NMR spectroscopies were used to compare the reactivity of GlbN and GlbN-A toward nitric oxide (NO). Both forms of the protein are capable of NO dioxygenase activity and both undergo heme bleaching after multiple NO challenges. GlbN and GlbN-A bind NO in the ferric state and form diamagnetic complexes (Fe
Sigma-Aldrich
Microvascular Endothelial Cell Basal Medium (500ml)