Merck
  • Activation of IGF-1/IGFBP-3 signaling by berberine improves intestinal mucosal barrier of rats with acute endotoxemia.
Activation of IGF-1/IGFBP-3 signaling by berberine improves intestinal mucosal barrier of rats with acute endotoxemia.
Fitoterapia (2017-11-21)
Yan He, Xiaoming Yuan, Guangrong Zhou, Aiwen Feng
ABSTRACT
Insulin-like growth factor I (IGF-I) and binding protein 3 (IGFBP-3) play a role in the maintenance of gut mucosal barrier function. Nevertheless, IGF-I/IGFBP-3 and tight junction protein (TJP) expression in small intestinal mucosa are often impaired during endotoxemia. In this model of acute endotoxemia, the regulatory effect of berberine on IGF-I/IGFBP-3 and TJP expression in ileal mucosa was evaluated. The findings revealed systemic injection of lipopolysaccharide (LPS) suppressed mRNA and protein expression of IGF-I and IGFBP-3, but berberine ameliorated their production. LPS injection inhibited occludin and claudin-1 protein generation, and this inhibitory effect of LPS was abolished by berberine. Inhibition of IGF-I/IGFBP-3 signaling by AG1024 or siRNAs reduced berberine-induced occludin and claudin-1 production. Additionally, GW9662 was found to repress berberine-induced IGF-I/IGFBP-3 expression, indicating of a cross-link between PPARγ and IGF-I/IGFBP-3 axis.
Sigma-Aldrich
MISSION® esiRNA, targeting human IGF1
Sigma-Aldrich
MISSION® esiRNA, targeting human IGFBP3