Merck
  • Pd(OH)2/C (Pearlman's catalyst): a highly active catalyst for Fukuyama, Sonogashira, and Suzuki coupling reactions.
Pd(OH)2/C (Pearlman's catalyst): a highly active catalyst for Fukuyama, Sonogashira, and Suzuki coupling reactions.
The Journal of organic chemistry (2003-02-15)
Yoshikazu Mori, Masahiko Seki
ABSTRACT
Treatment of thiol esters 1 with zinc reagent 2 in the presence of a small amount (<ca. 1 mol %) of nonpyrophoric Pd(OH)(2)/C (Pearlman's catalyst) provided functionalized asymmetrical ketones 3 in high yields. The use of Pd(OH)(2)/C was further applied to Sonogashira and Suzuki coupling reactions to afford the desired products in high yields.
Sigma-Aldrich
Palladium(II) chloride, ReagentPlus®, 99%
Sigma-Aldrich
Palladium(II) chloride, anhydrous, 60% Pd basis
Sigma-Aldrich
Palladium(II) chloride ChemBeads