Merck
  • Role of S100A3 in human hepatocellular carcinoma and the anticancer effect of sodium cantharidinate.
Role of S100A3 in human hepatocellular carcinoma and the anticancer effect of sodium cantharidinate.
Experimental and therapeutic medicine (2017-06-08)
Ran Tao, Zhong-Feng Wang, Wei Qiu, Yu-Fang He, Wei-Qun Yan, Wen-Yi Sun, Hai-Jun Li
ABSTRACT
The fifth most common cancer worldwide is hepatocellular carcinoma (HCC), which has an annual mortality rate of ~800,000. Although surgical procedures for HCC, such as hepatic resection and liver transplantation, have progressed and the outcomes of patients have improved, HCC is still characterized by frequent recurrence, even after liver transplantation. In the present study the expression of the protein coding gene, S100 calcium binding protein A3 (S100A3), was observed in 62 HCC tissues and tumor-surrounding tissues. The present study indicated that S100A3 activation was involved in tumorigenesis and tumor aggressiveness. The protein and mRNA expression levels of S100A3 in the human HCC cell line (HepG2) were investigated using western blotting and reverse transcription-quantitative polymerase chain reaction analysis, respectively. The function of sodium cantharidinate in inducing HCC cell apoptosis was also investigated. Sodium cantharidinate inhibited the protein and gene expression of S100A3 in HepG2 cells
Sigma-Aldrich
Monoclonal Anti-S100A3 antibody produced in mouse, clone 1D4, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Cantharidin