Merck
  • Enhanced long-chain fatty alcohol oxidation by immobilization of alcohol dehydrogenase from S. cerevisiae.
Enhanced long-chain fatty alcohol oxidation by immobilization of alcohol dehydrogenase from S. cerevisiae.
Applied microbiology and biotechnology (2017-11-02)
Carminna Ottone, Claudia Bernal, Néstor Serna, Andrés Illanes, Lorena Wilson
ABSTRACT
This work reports on the oxidation of long-chain aliphatic alcohols catalyzed by a stabilized alcohol dehydrogenase from S. cerevisiae (yeast alcohol dehydrogenase (YADH)). In particular, the oxidation of the fatty alcohol tetracosanol (C
Sigma-Aldrich
Lignoceryl alcohol, ≥99% (capillary GC)