- Exogenous DKK-3/REIC inhibits Wnt/β-catenin signaling and cell proliferation in human kidney cancer KPK1.
Exogenous DKK-3/REIC inhibits Wnt/β-catenin signaling and cell proliferation in human kidney cancer KPK1.
Oncology letters (2017-11-04)
Jiaqi Xu, Takuya Sadahira, Rie Kinoshita, Shun-Ai Li, Peng Huang, Koichiro Wada, Motoo Araki, Kazuhiko Ochiai, Hirofumi Noguchi, Masakiyo Sakaguchi, Yasutomo Nasu, Masami Watanabe
PMID29098038
ABSTRACT
The third member of the Dickkopf family (DKK-3), also known as reduced expression in immortalized cells (REIC), is a tumor suppressor present in a variety of tumor cells. Regarding the regulation of the Wnt/β-catenin signaling pathway, exogenous DKK-1 and DKK-2 are reported to inhibit Wnt signaling by binding the associated effectors. However, whether exogenous DKK-3 inhibits Wnt signaling remains unclear. A recombinant protein of human full-length DKK-3 was used to investigate the exogenous effects of the protein