Merck
  • Preventive effects of royal jelly against anaphylactic response in a murine model of cow's milk allergy.
Preventive effects of royal jelly against anaphylactic response in a murine model of cow's milk allergy.
Pharmaceutical biology (2017-10-07)
Malika Guendouz, Abir Haddi, Hadria Grar, Omar Kheroua, Djamel Saidi, Hanane Kaddouri
ABSTRACT
Royal jelly (RJ) has long been used to promote human health. The current study investigated the preventive effects of RJ against the development of a systemic and intestinal immune response in mice allergic to cow's milk proteins. Balb/c mice treated orally for seven days with RJ at doses of 0.5, 1 and 1.5 g/kg were sensitized intraperitoneally with β-lactoglobulin (β-Lg). Serum IgG and IgE anti-β-Lg were determined by an enzyme-linked immunosorbent assay (ELISA). Plasma histamine levels, symptom scores and body temperature were determined after in vivo challenge to β-Lg. Jejunums were used for assessment of local anaphylactic responses by an ex vivo study in Ussing chambers and morphologic changes by histological analysis. RJ significantly decreased serum IgG (31.15-43.78%) and IgE (64.28-66.6%) anti-β-Lg and effectively reduced plasma histamine level (66.62-67.36%) (p < 0.001) at all the doses tested. Additionally, no clinical symptoms or body temperature drops were observed in RJ-pretreated mice. Interestingly, RJ significantly reduced (p < 0.001) intestinal dysfunction by abolishing the secretory response (70.73-72.23%) induced by sensitization and prevented length aberrations of jejunal villi by 44.32-59.01% (p < 0.001). We speculate that using RJ may help prevent systemic and anaphylactic response in allergic mice. These effects may be related to its inhibitory effects on the degranulation of mast cells.
Sigma-Aldrich
Anti-Mouse IgG (Fc specific)–Biotin antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
ExtrAvidin®−Peroxidase, buffered aqueous solution