Merck
  • Spectrin and Other Membrane-Skeletal Components in Human Red Blood Cells of Different Age.
Spectrin and Other Membrane-Skeletal Components in Human Red Blood Cells of Different Age.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology (2017-07-03)
Annarita Ciana, Cesare Achilli, Giampaolo Minetti
ABSTRACT
Old human red blood cells (RBCs) have a reduced surface area with respect to young RBCs. If this decrease occurred through the release of vesicles similar to the spectrin-free vesicles that are shed in vitro under different experimental conditions or during storage, there would be no decrease of membrane-skeleton, but only of lipid bilayer surface area, during RBC ageing in vivo. However, we observed a decrease in spectrin and other membrane-skeletal proteins in old RBCs. Because RBCs contain components of the ubiquitin-proteasome system and other hydrolytic systems for protein degradation, we asked whether increased membrane-skeleton fragments could be detected in older RBCs. Four different anti-spectrin antibodies and an antibody anti-ubiquitin conjugates were used to analyse, by Western blotting, fragments of spectrin and other proteins in RBCs of different age separated in density gradients and characterized for their protein 4.1a/4.1b ratio as a cell age parameter. spectrin fragments do exist in RBCs of all ages, they represent a minute fraction of all spectrin, are membrane-bound and not cytoplasmic and do not increase with cell age. Besides spectrin, other membrane-skeletal components decrease with cell age. Observed results challenge the commonly accepted view that decrease in cell membrane throughout RBC life in vivo occurs via the release of spectrin-free vesicles.
Sigma-Aldrich
α-Cellulose, powder
Sigma-Aldrich
Sigmacell Cellulose, Type 50, 50 μm
Sigma-Aldrich
O-Acetyl-L-carnitine hydrochloride, ≥99% (titration), powder