Merck
  • Investigation of the surface plasmon polariton and electrochemical properties of covalent and non-covalent functionalized reduced graphene oxide.
Investigation of the surface plasmon polariton and electrochemical properties of covalent and non-covalent functionalized reduced graphene oxide.
Physical chemistry chemical physics : PCCP (2017-10-19)
Sanjit Saha, Pranab Samanta, Naresh Chandra Murmu, Tapas Kuila
ABSTRACT
The surface electronic properties of graphene oxide (GO) were modified through reduction and functionalization. Non-covalent functionalization was found to be superior compared to covalent functionalization due to the formation of few-layer graphene with a low defect content and average crystalline length. Because of the restoration of sp
Sigma-Aldrich
Sulfanilic acid azochromotrop, ≥80%