- Comparative metabolomics study on therapeutic mechanism of electro-acupuncture and moxibustion on rats with chronic atrophic gastritis (CAG).
Comparative metabolomics study on therapeutic mechanism of electro-acupuncture and moxibustion on rats with chronic atrophic gastritis (CAG).
Scientific reports (2017-11-01)
Cai-Chun Liu, Jiao-Long Chen, Xiao-Rong Chang, Qi-da He, Jia-Cheng Shen, Lin-Yu Lian, Ya-Dong Wang, Yuan Zhang, Fu-Qiang Ma, Hui-Ying Huang, Zong-Bao Yang
PMID29084954
ABSTRACT
Some studies have proved that both acupuncture and moxibustion are very effective for the treatment of CAG. However, little is known about therapeutic mechanism of electro-acupuncture and moxibustion on CAG as well as the difference between them. On the other hand, metabolomics is a 'top-down' approach to understand metabolic changes of organisms caused by disease or interventions in holistic context, which consists with the holistic thinking of electro-acupuncture and moxibustion treatment. In this study, the difference of therapeutic mechanism between electro-acupuncture and moxibustion on CAG rats was investigated by a