Merck
  • Retriever is a multiprotein complex for retromer-independent endosomal cargo recycling.
Retriever is a multiprotein complex for retromer-independent endosomal cargo recycling.
Nature cell biology (2017-09-12)
Kerrie E McNally, Rebecca Faulkner, Florian Steinberg, Matthew Gallon, Rajesh Ghai, David Pim, Paul Langton, Neil Pearson, Chris M Danson, Heike Nägele, Lindsey L Morris, Amika Singla, Brittany L Overlee, Kate J Heesom, Richard Sessions, Lawrence Banks, Brett M Collins, Imre Berger, Daniel D Billadeau, Ezra Burstein, Peter J Cullen
ABSTRACT
Following endocytosis into the endosomal network, integral membrane proteins undergo sorting for lysosomal degradation or are retrieved and recycled back to the cell surface. Here we describe the discovery of an ancient and conserved multiprotein complex that orchestrates cargo retrieval and recycling and, importantly, is biochemically and functionally distinct from the established retromer pathway. We have called this complex 'retriever'; it is a heterotrimer composed of DSCR3, C16orf62 and VPS29, and bears striking similarity to retromer. We establish that retriever associates with the cargo adaptor sorting nexin 17 (SNX17) and couples to CCC (CCDC93, CCDC22, COMMD) and WASH complexes to prevent lysosomal degradation and promote cell surface recycling of α5β1 integrin. Through quantitative proteomic analysis, we identify over 120 cell surface proteins, including numerous integrins, signalling receptors and solute transporters, that require SNX17-retriever to maintain their surface levels. Our identification of retriever establishes a major endosomal retrieval and recycling pathway.
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Roche
Anti-GFP, from mouse IgG1κ (clones 7.1 and 13.1)
Sigma-Aldrich
Anti-DSCR3 Antibody, from rabbit, purified by affinity chromatography