Merck
  • Cytokine-induced autophagy promotes long-term VCAM-1 but not ICAM-1 expression by degrading late-phase IκBα.
Cytokine-induced autophagy promotes long-term VCAM-1 but not ICAM-1 expression by degrading late-phase IκBα.
Scientific reports (2017-10-01)
Ling-Yun Chu, Ying-Chang Hsueh, Hsiao-Ling Cheng, Kenneth K Wu
ABSTRACT
Pro-inflammatory cytokines are known to induce endothelial cell autophagy, but the role of autophagy in regulating the expression of pro-inflammatory molecules has not been characterized. We hypothesized that autophagy facilitates expression of endothelial adhesion molecules. TNFα and IL-1β induced autophagy markers in human umbilical vein endothelial cells and inhibition of autophagy by 3-methyladenine (3-MA) blocked adhesion of Jurkat lymphocytes. Interestingly, 3-MA suppressed VCAM-1 but not ICAM-1 expression at 24 hours but not 6 hours. 3-MA suppressed VCAM-1 transcription and decreased nuclear NF-κB p65 level at 6 hours but not at 2 hours. Cytokines induced a biphasic degradation of IκBα and 3-MA selectively blocked the late-phase IκBα degradation. Our results suggest that cytokine-induced autophagy contributes to late-phase IκBα degradation, facilitates NF-κB nuclear translocation and VCAM-1 transcription for long-term VCAM-1 expression. With a cytokines array assay, we found that 3-MA also inhibited IP-10 expression. These findings provide new information about the role of endothelial autophagy in persistent expression of VCAM-1 and IP-10 which enhance lymphocyte recruitment and adhesion to endothelium.
Sigma-Aldrich
SB 202190, ≥98% (HPLC)
Sigma-Aldrich
3-Methyladenine, autophagy inhibitor
Sigma-Aldrich
NG25 trihydrochloride, ≥98% (HPLC)
Sigma-Aldrich
SP600125, ≥98% (HPLC)
Sigma-Aldrich
Bafilomycin A1 from Streptomyces griseus, ≥90% (HPLC)
Sigma-Aldrich
PD 98,059, solid
Sigma-Aldrich
Rapamycin from Streptomyces hygroscopicus, ≥95% (HPLC), powder
Sigma-Aldrich
Bay 11-7082, ≥98% (HPLC), powder
Sigma-Aldrich
MG-132(R), ≥95% (HPLC)