Merck
Interaction of [V
Chemistry, an Asian journal (2017-06-27)
Isabel Correia, Ielyzaveta Chorna, Isabel Cavaco, Somnath Roy, Maxim L Kuznetsov, Nádia Ribeiro, Gonçalo Justino, Fernanda Marques, Teresa Santos-Silva, Marino F A Santos, Hugo M Santos, José L Capelo, James Doutch, João Costa Pessoa
ABSTRACT
[VO(acac)
Sigma-Aldrich
Albumin from human serum, lyophilized powder, Fatty acid free, Globulin free, ≥99% (agarose gel electrophoresis)
Pricing and availability is not currently available.