Merck
  • A novel Salmonella strain inactivated by a regulated autolysis system and expressing the B subunit of Shiga toxin 2e efficiently elicits immune responses and confers protection against virulent Stx2e-producing Escherichia coli.
A novel Salmonella strain inactivated by a regulated autolysis system and expressing the B subunit of Shiga toxin 2e efficiently elicits immune responses and confers protection against virulent Stx2e-producing Escherichia coli.
BMC veterinary research (2017-02-02)
Gayeon Won, Tae Hoon Kim, John Hwa Lee
ABSTRACT
Salmonella Typhimurium (S. Typhimurium) inactivated by a regulated autolysis system was genetically engineered to express the homo-pentameric B subunit of Shiga toxin 2e (Stx2eB) on its surface. To prepare a strain able to yield autolyzed Salmonella bearing Stx2eB, the plasmid pJHL184 harboring stx JOL1454 provided superior immunogenicity and effective protection against challenge with a sublethal dose, which demonstrates its potential as a candidate vaccine against edema disease.
Sigma-Aldrich
1,3-Diaminopropane, ≥99%