Merck
  • Rab11-FIP3 Regulation of Lck Endosomal Traffic Controls TCR Signal Transduction.
Rab11-FIP3 Regulation of Lck Endosomal Traffic Controls TCR Signal Transduction.
Journal of immunology (Baltimore, Md. : 1950) (2017-02-27)
Jérôme Bouchet, Iratxe Del Río-Iñiguez, Elena Vázquez-Chávez, Rémi Lasserre, Sonia Agüera-González, Céline Cuche, Mary W McCaffrey, Vincenzo Di Bartolo, Andrés Alcover
ABSTRACT
The role of endosomes in receptor signal transduction is a long-standing question, which remains largely unanswered. The T cell Ag receptor and various components of its proximal signaling machinery are associated with distinct endosomal compartments, but how endosomal traffic affects T cell signaling remains ill-defined. In this article, we demonstrate in human T cells that the subcellular localization and function of the protein tyrosine kinase Lck depends on the Rab11 effector FIP3 (Rab11 family interacting protein-3). FIP3 overexpression or silencing and its ability to interact with Rab11 modify Lck subcellular localization and its delivery to the immunological synapse. Importantly, FIP3-dependent Lck localization controls early TCR signaling events, such as tyrosine phosphorylation of TCRζ, ZAP70, and LAT and intracellular calcium concentration, as well as IL-2 gene expression. Interestingly, FIP3 controls both steady-state and poststimulation phosphotyrosine and calcium levels. Finally, our findings indicate that FIP3 modulates TCR-CD3 cell surface expression via the regulation of steady-state Lck-mediated TCRζ phosphorylation, which in turn controls TCRζ protein levels. This may influence long-term T cell activation in response to TCR-CD3 stimulation. Therefore, our data underscore the importance of finely regulated endosomal traffic in TCR signal transduction and T cell activation leading to IL-2 production.
Sigma-Aldrich
MISSION® esiRNA, targeting human IKBKG