Merck
  • Aggregation of a hepatitis C virus replicase module induced by ablation of p97/VCP.
Aggregation of a hepatitis C virus replicase module induced by ablation of p97/VCP.
The Journal of general virology (2017-07-12)
Zhigang Yi, Zhenghong Yuan
ABSTRACT
Hijacking host membranes to assemble a membrane-associated viral replicase is a hallmark of almost all positive-strand RNA viruses. However, how the virus co-opts host factors to facilitate this energy-unfavourable process is incompletely understood. In a previous study, using hepatitis C virus (HCV) as a model and employing affinity purification of the viral replicase, we identified a valosin-containing protein (p97/VCP), a member of the ATPases associated with diverse cellular activities (AAA+ ATPase family), as a viral replicase-associated host factor. It is required for viral replication, depending on its ATPase activity. In this study, we used VCP pharmacological inhibitors and short hairpin (sh) RNA-mediated knockdown to ablate VCP function and then dissected the roles of VCP in viral replicase assembly in an HCV subgenomic replicon system and a viral replicase assembly surrogate system. Ablation of VCP specifically resulted in the pronounced formation of an SDS-resistant aggregation of HCV NS5A and the reduction of hyperphosphorylation of NS5A. The NS5A dimerization domain was indispensable for aggregation and the NS5A disordered regions also contributed to a lesser extent. The reduction of the hyperphosphorylation of NS5A coincided with the aggregation of NS5A. We propose that HCV may co-opt VCP to disaggregate an aggregation-prone replicase module to facilitate its replicase assembly.
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
DBeQ, ≥98% (HPLC)
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)