Merck
  • Different genetic structures revealed resident populations of a specialist parasitoid wasp in contrast to its migratory host.
Different genetic structures revealed resident populations of a specialist parasitoid wasp in contrast to its migratory host.
Ecology and evolution (2017-08-05)
Shu-Jun Wei, Yuan Zhou, Xu-Lei Fan, Ary A Hoffmann, Li-Jun Cao, Xue-Xin Chen, Zai-Fu Xu
ABSTRACT
Genetic comparisons of parasitoids and their hosts are expected to reflect ecological and evolutionary processes that influence the interactions between species. The parasitoid wasp,