Merck
  • Thermoresponsive nanocomposite gel for local drug delivery to suppress the growth of glioma by inducing autophagy.
Thermoresponsive nanocomposite gel for local drug delivery to suppress the growth of glioma by inducing autophagy.
Autophagy (2017-06-09)
Li Ding, Qi Wang, Ming Shen, Ying Sun, Xiangyu Zhang, Can Huang, Jianhua Chen, Rongxin Li, Yourong Duan
ABSTRACT
Although the treatments of malignant glioma include surgery, radiotherapy and chemotherapy by oral drug administration, the prognosis of patients with glioma remains very poor. We developed a polyethylene glycol-dipalmitoylphosphatidyle- thanoiamine (mPEG-DPPE) calcium phosphate nanoparticles (NPs) injectable thermoresponsive hydrogel (nanocomposite gel) that could provide a sustained and local delivery of paclitaxel (PTX) and temozolomide (TMZ). In addition, the proportion of PTX and TMZ for the optimal synergistic antiglioma effect on C6 cells was determined to be 1:100 (w/w) by the Chou and Talalay method. Our results clearly indicated that the autophagy induced by PTX:TMZ NPs plays an important role in regulating tumor cell death, while autophagy inhibition dramatically reverses the antitumor effect of PTX:TMZ NPs, suggesting that antiproliferative autophagy occurs in response to PTX:TMZ NPs treatment. The antitumor efficacy of the PTX:TMZ NP-loaded gel was evaluated in situ using C6 tumor-bearing rats, and the PTX:TMZ NP-loaded gel exhibited superior antitumor performance. The antitumor effects of the nanocomposite gel in vivo were shown to correlate with autophagic cell death in this study. The in vivo results further confirmed the advantages of such a strategy. The present study may provide evidence supporting the development of nanomedicine for potential clinical application.
Sigma-Aldrich
Temozolomide, ≥98% (HPLC)
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid
Sigma-Aldrich
Annexin V-FITC Apoptosis Detection Kit
Sigma-Aldrich
Dansylcadaverine, BioReagent, suitable for fluorescence, ≥99.0% (HPLC)
Sigma-Aldrich
Alginic acid sodium salt, powder