Merck
  • Sequence of the N-terminal half of Bacillus amyloliquefaciens alpha-amylase.
Sequence of the N-terminal half of Bacillus amyloliquefaciens alpha-amylase.
The Biochemical journal (1980-02-01)
H Chung, F Friedberg
ABSTRACT
Bacillus amyloliquefaciens alpha-amylase (1,4-alpha-D-glucan glucanohydrolase. EC 3.2.1.1), which is commercially supplied as 'Bacillus subtilis alpha-amylase' does not cross-react immunologically with B. subtilis alpha-amylase. This enzyme (from B. amyloliquefaciens) was cleaved by treatment with CNBr into seven fragments. Peptide A was selected for sequence determination. It is the longest one, containing 185 amino acids (i.e. approx. 50% of the total molecule) and connects to the hexapeptide of the N-terminus. Its primary structure was aligned by use of various proteolytic enzymes. The sequence of amino acids 181-184 is identical with that of amino acids 14-17 of the alpha-amylase isolated from B. subtilis (except that amino acid 183 is asparagine rather than aspartic acid).
Sigma-Aldrich
α-Amylase from Bacillus sp., powder, ≥400 units/mg protein (Lowry)
Sigma-Aldrich
α-Amylase from Bacillus sp., Type II-A, lyophilized powder, ≥1,500 units/mg protein (biuret)