Merck
Flexibility and Stability of Metal Coordination Macromolecules.
Chemistry (Weinheim an der Bergstrasse, Germany) (2017-03-24)
Heyan Jiang, Diya Geng, Dapeng Liu, Nicholas Lanigan, Xiaosong Wang
ABSTRACT
The effect of chain structure on flexibility and stability of macromolecules containing weak P-Fe metal coordination bonds is studied. Migration insertion polymerization (MIP) of FpC
Sigma-Aldrich
1,3-Bis(diphenylphosphino)propane, 97%
Sigma-Aldrich
1,6-Dichlorohexane, 98%