Merck
  • Molecular interactions of EphA4, growth hormone receptor, Janus kinase 2, and signal transducer and activator of transcription 5B.
Molecular interactions of EphA4, growth hormone receptor, Janus kinase 2, and signal transducer and activator of transcription 5B.
PloS one (2017-07-08)
Takahiro Sawada, Daiki Arai, Xuefeng Jing, Masayasu Miyajima, Stuart J Frank, Kazushige Sakaguchi
ABSTRACT
We previously reported that EphA4, a member of the Eph family of receptor tyrosine kinases, is an important modulator of growth hormone (GH) signaling, leading to augmented synthesis of insulin-like growth factor 1 (IGF1) for postnatal body growth. In the present study, we report the molecular interactions of EphA4, GH receptor (GHR), Janus kinase 2 (JAK2), and signal transducer and activator of transcription 5B (STAT5B). EphA4 binds to GHR at both its extracellular and intracellular domains and phosphorylates GHR when stimulated with a ligand. The cytoplasmic domain of EphA4 binds to the carboxy-terminus of JAK2 in contrast to the known binding of GHR to the amino-terminus. STAT5B binds to the amino-terminal kinase domain of EphA4. Ligand-activated EphA4 and JAK2 phosphorylate each other and STAT5B, but JAK2 does not appear to phosphorylate EphA4-bound STAT5B. Ligand-activated EphA4 induces the nuclear translocation of STAT5B in a JAK2-independent manner. GHR expression is required for the activation of STAT5B signaling, even via the JAK2-independent pathway. Various ephrins that have affinity for EphA4 induce STAT5B phosphorylation. These findings suggest the molecular mechanisms by which ephrin/EphA4 signaling enhances the canonical GH-IGF1 axis.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Ephrin-A5/Fc Chimera human, >90% (SDS-PAGE), recombinant, expressed in NSO cells, lyophilized powder
Sigma-Aldrich
Ephrin-B2/Fc Chimera from mouse, >90% (SDS-PAGE), recombinant, expressed in NSO cells, lyophilized powder
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium - high glucose, With 4500 mg/L glucose, L-glutamine, and sodium bicarbonate, without sodium pyruvate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Anti-Phosphotyrosine Antibody, clone 4G10®, clone 4G10®, Upstate®, from mouse
Sigma-Aldrich
Phosphate buffered saline, BioPerformance Certified, pH 7.4
Sigma-Aldrich
Somatotropin from human pituitary, recombinant, expressed in E. coli, ≥95% (SDS-PAGE), lyophilized powder
Sigma-Aldrich
Ephrin-A1/Fc Chimera from mouse, >90% (SDS-PAGE), recombinant, expressed in NSO cells, lyophilized powder
Sigma-Aldrich
Minimum Essential Medium Eagle, With Earle′s salts, L-glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture