- Immune hyperreactivity of Aβ plaque-associated microglia in Alzheimer's disease.
Immune hyperreactivity of Aβ plaque-associated microglia in Alzheimer's disease.
Neurobiology of aging (2017-04-25)
Zhuoran Yin, Divya Raj, Nasrin Saiepour, Debby Van Dam, Nieske Brouwer, Inge R Holtman, Bart J L Eggen, Thomas Möller, Joseph A Tamm, Aicha Abdourahman, Elly M Hol, Willem Kamphuis, Thomas A Bayer, Peter P De Deyn, Erik Boddeke
PMID28434692
ABSTRACT
Alzheimer's disease (AD) is strongly associated with microglia-induced neuroinflammation. Particularly, Aβ plaque-associated microglia take on an "activated" morphology. However, the function and phenotype of these Aβ plaque-associated microglia are not well understood. We show hyperreactivity of Aβ plaque-associated microglia upon systemic inflammation in transgenic AD mouse models (i.e., 5XFAD and APP23). Gene expression profiling of Aβ plaque-associated microglia (major histocompatibility complex II
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O111:B4, γ-irradiated, BioXtra, suitable for cell culture