Merck
  • Nanosecond laser ablation enhances cellular infiltration in a hybrid tissue scaffold.
Nanosecond laser ablation enhances cellular infiltration in a hybrid tissue scaffold.
Materials science & engineering. C, Materials for biological applications (2017-05-24)
S Jahnavi, N Arthi, S Pallavi, C Selvaraju, G S Bhuvaneshwar, T V Kumary, R S Verma
ABSTRACT
Hybrid tissue engineered (HTE) scaffolds constituting polymeric nanofibers and biological tissues have attractive bio-mechanical properties. However, they suffer from small pore size due to dense overlapping nanofibers resulting in poor cellular infiltration. In this study, using nanosecond (ns) laser, we fabricated micro-scale features on Polycaprolactone (PCL)-Chitosan (CH) nanofiber layered bovine pericardium based Bio-Hybrid scaffold to achieve enhanced cellular adhesion and infiltration. The laser energy parameters such as fluence of 25J/cm
Sigma-Aldrich
Collagenase from Clostridium histolyticum, suitable for release of physiologically active rat epididymal adipocytes, Type II, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid
Sigma-Aldrich
Ribonuclease A from bovine pancreas, for molecular biology, ≥70 Kunitz units/mg protein, lyophilized
Sigma-Aldrich
Chitosan, from shrimp shells, practical grade
Sigma-Aldrich
Polycaprolactone, average Mn 80,000
Sigma-Aldrich
Deoxyribonuclease I from bovine pancreas, Type II, lyophilized powder, Protein ≥80 %, ≥2,000 units/mg protein
Sigma-Aldrich
Proteinase K from Tritirachium album, lyophilized powder, BioUltra, ≥30 units/mg protein, for molecular biology