- New Ether-functionalized Morpholinium- and Piperidinium-based Ionic Liquids as Electrolyte Components in Lithium and Lithium-Ion Batteries.
New Ether-functionalized Morpholinium- and Piperidinium-based Ionic Liquids as Electrolyte Components in Lithium and Lithium-Ion Batteries.
ChemSusChem (2017-04-14)
Maria Assunta Navarra, Kanae Fujimura, Mirko Sgambetterra, Akiko Tsurumaki, Stefania Panero, Nobuhumi Nakamura, Hiroyuki Ohno, Bruno Scrosati
PMID28407378
ABSTRACT
Here, two ionic liquids, N-ethoxyethyl-N-methylmorpholinium bis(trifluoromethanesulfonyl)imide (M